EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Validation-based insertional mutagenesis identifies lysine demethylase FBXL11 as a negative regulator of NFkappaB.
PMID 19805303
Abstract
We describe a highly efficient use of lentiviral validation-based insertional mutagenesis (VBIM) to generate large populations of mammalian cells in which a strong promoter is inserted into many different genomic loci, causing greatly increased expression of downstream sequences. Many different selections or screens can follow, to isolate dominant mutant clones with a desired phenotypic change. The inserted promoter can be excised or silenced at will, to prove that the insertion caused the mutation. Cloning DNA flanking the insertion site identifies the locus precisely. VBIM virus particles are pseudotyped with VSV G protein, allowing efficient infection of most mammalian cell types, including non-dividing cells, and features are included that give high yields of stable virus stocks. In several different selections, useful mutants have been obtained at frequencies of approximately 10(-6) or higher. We used the VBIM technique to isolate mutant human cells in which the F-box leucine-rich protein 11 (FBXL11), a histone H3K36 demethylase, is shown to be a negative regulator of NFkappaB. High levels of FBXL11 block the ability of NFkappaB to bind to DNA or activate gene expression, and siRNA-mediated reduction of FBXL11 expression has the opposite effects. The H212A mutation of FBXL11 abolishes both its histone H3K36 demethylase activity and its ability to inhibit NFkappaB. Thus, we have used a powerful tool for mutagenesis of mammalian cells to reveal an aspect of the complex regulation of NFkappaB-dependent signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV31697
Anti-FBXL11 antibody produced in rabbit, affinity isolated antibody