EMAIL THIS PAGE TO A FRIEND
Natural product research
Chemical constituents from the fruits of Hippophae rhamnoides.
PMID 19809919
Abstract
Ten compounds were isolated from the fruits of Hippophae rhamnoides. On the basis of spectroscopic and chemical methods, the structures of these compounds were elucidated as hippophae cerebroside (1), oleanolic acid (2), ursolic acid (3), 19-alpha-hydroxyursolic acid (4), dulcioic acid (5), 5-hydroxymethyl-2-furancarbox-aldehyde (6), cirsiumaldehyde (7), octacosanoic acid (8), palmitic acid (9) and 1-O-hexadecanolenin (10). Among them, 1 was a new compound, and 4-7 and 10 were obtained from the genus for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284432
Octacosanoic acid, synthetic, ≥98%
C28H56O2