EMAIL THIS PAGE TO A FRIEND
The European journal of neuroscience
IQ-ArfGEF/BRAG1 is associated with synaptic ribbons in the mouse retina.
PMID 19811534
Abstract
IQ-ArfGEF/BRAG1 is a guanine nucleotide exchange factor for ADP ribosylation factors (Arfs), which are implicated in membrane trafficking and actin cytoskeleton dynamics. In this study, we examined the immunohistochemical localization of IQ-ArfGEF/BRAG1 in the adult mouse retina using light and electron microscopy. IQ-ArfGEF/BRAG1 was distributed in a punctate manner and colocalized well with RIBEYE in both the outer and inner plexiform layers. Immunoelectron microscopic analysis showed that IQ-ArfGEF/BRAG1 was localized at the synaptic ribbons of photoreceptors. When heterologously expressed in HeLa cells, IQ-ArfGEF/BRAG1 was recruited to RIBEYE-containing clusters and formed an immunoprecipitable complex with RIBEYE. Furthermore, immunoprecipitation analysis showed that anti-IQ-ArfGEF/BRAG1 antibody efficiently pulled down RIBEYE from retinal lysates. These findings indicate that IQ-ArfGEF/BRAG1 is a novel component of retinal synaptic ribbons and forms a protein complex with RIBEYE.