EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Comparison of photodegradative efficiencies and mechanisms of Victoria Blue R assisted by Nafion-coated and fluorinated TiO2 photocatalysts.
PMID 19815344
Abstract
The purposes of this research were to study the effects of two modified photocatalysts, Nafion-coated TiO(2) and fluorinated TiO(2), and photocatalytic degradation of Victoria Blue R in aqueous solution. Photocatalytic degradation of Victoria Blue R was accelerated by the modified photocatalysts. Bulk and surface characterizations of the resulting powders were carried out. Attachment of the anions to the TiO(2) surface using the Nafion-coated-TiO(2) possibly results in increased adsorption of the cationic dye, and the degradation rate is larger for the cationic dye. It was found that Victoria Blue R on the two illuminated TiO(2) surfaces underwent very different changes. To obtain a better understanding on the mechanistic details of this modified-TiO(2)-assisted photodegradation of the Victoria Blue R dye with UV irradiation, a large number of intermediates of the process were separated, identified, and characterized by a high-performance liquid chromatography-mass spectrometry technique. Several probable photodegradation pathways were proposed and discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
234176
Victoria Blue R, Dye content 80 %
C29H32ClN3