EMAIL THIS PAGE TO A FRIEND
Current Protocols in Cytometry
High-resolution multiparameter DNA flow cytometry for the detection and sorting of tumor and stromal subpopulations from paraffin-embedded tissues.
PMID 19816924