EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Ultrafast relaxation dynamics of the excited states of 1-amino- and 1-(N,N-dimethylamino)-fluoren-9-ones.
PMID 19821474
Abstract
The dynamics of the excited states of 1-aminofluoren-9-one (1AF) and 1-(N,N-dimethylamino)-fluoren-9-one (1DMAF) are investigated by using steady-state absorption and fluorescence as well as subpicosecond time-resolved absorption spectroscopic techniques. Following photoexcitation of 1AF, which exists in the intramolecular hydrogen-bonded form in aprotic solvents, the excited-state intramolecular proton-transfer reaction is the only relaxation process observed in the excited singlet (S(1)) state. However, in protic solvents, the intramolecular hydrogen bond is disrupted in the excited state and an intermolecular hydrogen bond is formed with the solvent leading to reorganization of the hydrogen-bond network structure of the solvent. The latter takes place in the timescale of the process of solvation dynamics. In the case of 1DMAF, the main relaxation pathway for the locally excited singlet, S(1)(LE), or S(1)(ICT) state is the configurational relaxation, via nearly barrierless twisting of the dimethylamino group to form the twisted intramolecular charge-transfer, S(1)(TICT), state. A crossing between the excited-state and ground-state potential energy curves is responsible for the fast, radiationless deactivation and nonemissive character of the S(1)(TICT) state in polar solvents, both aprotic and protic. However, in viscous but strong hydrogen-bond-donating solvents, such as ethylene glycol and glycerol, crossing between the potential energy surfaces for the ground electronic state and the hydrogen-bonded complex formed between the S(1)(TICT) state and the solvent is possibly avoided and the hydrogen-bonded complex is weakly emissive.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F1506
9-Fluorenone, 98%
C13H8O