EMAIL THIS PAGE TO A FRIEND
European journal of dermatology : EJD
Contact immunotherapy-induced Renbök phenomenon in a patient with alopecia areata and psoriasis vulgaris.
PMID 19822483
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
339792
3,4-Dibutoxy-3-cyclobutene-1,2-dione, 98%
C12H18O4