EMAIL THIS PAGE TO A FRIEND
Dental materials journal
Bonding durability against water of a fluorine-containing resin for precious metal alloys.
PMID 19822997
Abstract
In order to clarify the effect of fluorination of an adhesive resin on the durability of the resin bond to precious metal alloys, 2,2,2-trifluoroethyl methacrylate (TFEMA)-poly(2,2,2-trifluoroethyl methacrylate) (PTFEMA)/TBBO adhesive resin was prepared. The tensile bond strength of this resin to precious metal alloys treated with 9,10-epithiodecyl methacrylate (EP8MA) generally increased in the order Ag alloy
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
373761
2,2,2-Trifluoroethyl methacrylate, contains 50-200 ppm MEHQ as inhibitor, 99%
C6H7F3O2