EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis of new sugar derivatives and evaluation of their antibacterial activities against Mycobacterium tuberculosis.
PMID 19828313
Abstract
A series of sugar derivatives (1-13) were synthesized and evaluated for antibacterial activity against Mycobacteriumtuberculosis (MTB), especially multi-drug resistant (MDR) MTB, and the structure-activity relationships of these compounds were studied. The results showed that the compound OCT313 (2-acetamido-2deoxy-beta-D-glucopyranosyl N,N-dimethyldithiocarbamate) (4) exhibited significant in vitro bactericidal activity, and that the dithiocarbamate group at C-1 position of the glucopyranoside ring was requisite for the antibacterial activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
126365
Dimethylamine hydrochloride, 99%
C2H7N · HCl
38960
Dimethylamine hydrochloride, purum, ≥98.0% (AT)
C2H7N · HCl