EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Efficient method for the synthesis of functionalized pyrazoles by catalyst-free one-pot tandem reaction of nitroalkenes with ethyl diazoacetate.
PMID 19830281
Abstract
The one-pot synthesis of multisubstituted pyrazole derivatives was achieved via catalyst-free 1,3-dipolar cycloaddition of ethyl diazoacetate and nitroalkenes as the key step and elimination of the leaving group (NO(2) or Br) followed by intramolecular proton transfer with satisfactory yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E22201
Ethyl diazoacetate, contains ≥13 wt. % dichloromethane
C4H6N2O2