EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Macrocyclic oligoureas with xanthene and diphenyl ether units.
PMID 19830287
Abstract
Two cyclic oligoureas with 64- and 80-membered rings in which two sets of three or four rigid xanthene (X) units are connected via flexible diphenyl ether (D) units were synthesized by a stepwise fragment condensation. The compounds were characterized by (1)H NMR and ESI mass spectrometry. The structure of the cyclic octamer (XXXDXXXD) was additionally confirmed by single crystal X-ray analysis. The molecule assumes a strongly folded conformation with distorted C(2)-symmetry, stabilized by intramolecular hydrogen bonds. Surprisingly, intermolecular hydrogen bonds between the macrocycles were not observed. (1)H NMR spectra suggest a C(2) symmetrical conformation of the octamer in solution also, while its kinetically stable complex with three chloride ions has no symmetry element at all.