EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Estimation of antioxidant capacity against pathophysiologically relevant oxidants using Pyrogallol Red.
PMID 19833093
Abstract
Peroxynitrite and hypochlorite are oxidants relevant in many pathological situations. We propose a simple spectrophotometric assay to determine antioxidant capacity against hypochlorite and peroxynitrite based on protection against Pyrogallol Red decolorization. The assay can be performed on a microplate and requires minute amounts of material. Standard antioxidants show different reactivities for both oxidants. Antioxidant capacity of blood plasma (anticoagulated with EDTA) of healthy persons was found to be 559+/-49 micromol/l and 11.6+/-1.2 mmol/l of ascorbic acid equivalents for peroxynitrite and hypochlorite, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8759
Pyrogallol Red
C19H12O8S