EMAIL THIS PAGE TO A FRIEND
Journal of Biological Engineering
Fluorometric determination of ethidium bromide efflux kinetics in Escherichia coli.
PMID 19835592
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AG100 Human IgG Fc Control Protein, recombinant protein