EMAIL THIS PAGE TO A FRIEND
Steroids
An efficient, practical synthesis of 2-methoxyestradiol.
PMID 19835898
Abstract
An efficient and practical scheme to synthesize 2-methoxyestradiol has been developed. The key step was the copper-mediated methoxylation using ethyl acetate as a co-catalyst to introduce a methoxyl group. These synthetic procedures of four steps from 17beta-estradiol as starting material gave 2-methoxyestradiol with a 61% overall yield.