EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Oxiranylmethyloxy or thiiranylmethyloxy-azaxanthones and -acridone analogues as potential topoisomerase I inhibitors.
PMID 19836231
Abstract
A total of seven new oxyranylmethyloxy or thiiranylmethyloxy group substituted 5-azaxanthones and -acridones analogues were synthesized and tested for their biological activities for cancer cell lines and topoisomerases. Among the compounds, compound 5, 3-thiiranylmethyloxy-1-hydroxy-5-azaxanthone, showed effective topoisomerase I inhibitory activity, 50% and 27% inhibition ratio at 100 and 20muM, respectively. This result is the first finding of the function of 5-azaxanthone compounds for topoisomerase I inhibition and can provide a novel skeleton for the anticancer drug development process.