EMAIL THIS PAGE TO A FRIEND
Clinical biochemistry
Identification of a neonate with hepatorenal tyrosinemia by combined routine newborn screening for succinylacetone, acylcarnitines and amino acids.
PMID 19836366
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1415
4,6-Dioxoheptanoic acid, powder
C7H10O4