EMAIL THIS PAGE TO A FRIEND
Nature genetics
Mutations in FAM134B, encoding a newly identified Golgi protein, cause severe sensory and autonomic neuropathy.
PMID 19838196
Abstract
Hereditary sensory and autonomic neuropathy type II (HSAN II) leads to severe mutilations because of impaired nociception and autonomic dysfunction. Here we show that loss-of-function mutations in FAM134B, encoding a newly identified cis-Golgi protein, cause HSAN II. Fam134b knockdown results in structural alterations of the cis-Golgi compartment and induces apoptosis in some primary dorsal root ganglion neurons. This implicates FAM134B as critical in long-term survival of nociceptive and autonomic ganglion neurons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV44827
Anti-FAM134B antibody produced in rabbit, IgG fraction of antiserum