EMAIL THIS PAGE TO A FRIEND
Oncogene
DNA damage induces Chk1-dependent threonine-160 phosphorylation and activation of Cdk2.
PMID 19838212
Abstract
Abnormal centrosome numbers arise in tumours and can cause multipolar mitoses and genome instability. Cdk2 controls normal centrosome duplication, but Chk1-dependent centrosome amplification also occurs after DNA damage. We investigated the involvement of cyclin-dependent kinases (Cdks) in DNA damage-induced centrosome amplification using cells lacking either Cdk2, or both Cdk1 and Cdk2 activity. Cdk2(-/-) DT40 cells showed robust centrosome amplification after ionizing radiation (IR), whereas Cdk1-deficient Cdk2(-/-) cells showed no centrosome amplification, demonstrating that Cdk1 can substitute for Cdk2 in this pathway. Surprisingly, we found that Cdk2 activity was upregulated by IR in wild-type but not in Chk1(-/-) DT40 cells. Cdk2 upregulation also occurred in HeLa cells after IR treatment. Chk1-dependent Cdk2 induction was not accompanied by increased levels of Cdk1, Cdk2, cyclin A or cyclin E, but activating T160 phosphorylation of Cdk2 increased after IR. Moreover, Cdk2 overexpression restored IR-induced centrosome amplification in Cdk1-deficient Cdk2(-/-) cells, but T160A mutation blocked this rescue. Our data suggest that Chk1 signalling causes centrosome amplification after IR by upregulating Cdk2 activity through activating phosphorylation.
Related Materials