EMAIL THIS PAGE TO A FRIEND
ChemSusChem
The derivatization of bioplatform molecules by using KF/alumina catalysis.
PMID 19842156
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I29204
Itaconic acid, ≥99%
C5H6O4