EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Deconvolution of complex NMR spectra in small molecules by multi frequency homonuclear decoupling (MDEC).
PMID 19845379
Abstract
A new technique to deconvolute complex (1)H NMR spectra of small molecules has been developed that utilizes shape selective pulses to simultaneously decouple multiple protons. A limitation in the assignment of the relative configuration of small molecules is the ability to accurately obtain coupling constants. Other methods such as the E.COSY and the 2D J-resolved are available to obtain complicated coupling constants; the multiple homonuclear decoupling method (MDEC) described is a rapid and simple technique. Three examples of increasing spectral complexity, menthol, cholesteryl acetate and a C(16) fatty acid, demonstrate the utility of the technique. Increasing the experimental utility, the single pulse MDEC experiment can be incorporated in other 1D experiments, such as a 1D-TOCSY to solve specific problems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
151114
Cholesteryl acetate, 97%
C29H48O2