EMAIL THIS PAGE TO A FRIEND
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists
Homotaurine: a failed drug for Alzheimer's disease and now a nutraceutical for memory protection.
PMID 19850791
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A76109
3-Amino-1-propanesulfonic acid, 97%
C3H9NO3S