EMAIL THIS PAGE TO A FRIEND
Circulation journal : official journal of the Japanese Circulation Society
Mitochondrial protection and the reversal of left ventricular remodeling.
PMID 19851030
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0742 EUK-8, ≥95% (HPLC)
C16H14ClMnN2O2