EMAIL THIS PAGE TO A FRIEND
Natural product research
A chromanone alkaloid from Derris ovalifolia stem.
PMID 19851932
Abstract
From the stem of Derris ovalifolia, a new chromanone alkaloid characterised as 7-methoxy-3-[1-(3-pyridyl)methylidene]-4-chromanone (1) was isolated along with four known compounds, dehydrotoxicarol (2), dehydrodeguelin (3), toxicarol isoflavone (4) and indole-3-carbaldehyde (5). Their structures were determined on the basis of spectral data interpretation. The structure of compound 1 has also been confirmed by synthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
122351
4-Chromanone, 97%
C9H8O2
129445
Indole-3-carboxaldehyde, 97%
C9H7NO