EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
Differential effects of reactive nitrogen species on DNA base excision repair initiated by the alkyladenine DNA glycosylase.
PMID 19864471
Abstract
Chronic generation of reactive nitrogen species (RNS) can cause DNA damage and may also directly modify DNA repair proteins. RNS-modified DNA is repaired predominantly by the base excision repair (BER) pathway, which includes the alkyladenine DNA glycosylase (AAG). The AAG active site contains several tyrosines and cysteines that are potential sites for modification by RNS. In vitro, we demonstrate that RNS differentially alter AAG activity depending on the site and type of modification. Nitration of tyrosine 162 impaired 1,N(6)-ethenoadenine (epsilonA)-excision activity, whereas nitrosation of cysteine 167 increased epsilonA excision. To understand the effects of RNS on BER in vivo, we examined intestinal adenomas for levels of inducible nitric oxide synthase (iNOS) and AAG. A striking correlation between AAG and iNOS expression was observed (r = 0.76, P = 0.00002). Interestingly, there was no correlation between changes in AAG levels and enzymatic activity. We found AAG to be nitrated in human adenomas, suggesting that this RNS modification is relevant in the human disease. Expression of key downstream components of BER, apurinic/apyrimidinic endonuclease 1 (APE1) and DNA polymerase beta (POLbeta), was also examined. POLbeta protein was increased in nearly all adenomas compared with adjacent non-tumor tissues, whereas APE1 expression was only increased in approximately half of the adenomas and also was relocalized to the cytoplasm in adenomas. Collectively, the results suggest that BER is dysregulated in colon adenomas. RNS-induced posttranslational modification of AAG is one mechanism of BER dysregulation, and the type of modification may define the role of AAG during carcinogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-284
Anti-Nitrotyrosine Antibody