EMAIL THIS PAGE TO A FRIEND
Pediatrics
Methsuximide for intractable childhood seizures.
PMID 1987529
Abstract
Methsuximide was added to the therapeutic regimens of 25 children with intractable epilepsy. In 15 patients the drug was well tolerated and resulted in a 50% or greater reduction in seizure frequency. No serious or irreversible adverse effects were seen. Methsuximide is frequently overlooked and may be an effective adjunctive antiepileptic for children with intractable seizures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
860581
α-Methyl-α-phenylsuccinimide, 99%
C11H11NO2