EMAIL THIS PAGE TO A FRIEND
Magnetic resonance in chemistry : MRC
New oxidized sterols from Aspergillus awamori and the endo-boat conformation adopted by the cyclohexene oxide system.
PMID 19877128
Abstract
Two new oxidized sterols 1 and 2 were obtained from the active fraction of a mangrove fungus Aspergillus awamori isolated from the soils around the mangrove plant Acrostichum speciosum. Their structures were elucidated using spectroscopic methods as 22E-7alpha-methoxy-5alpha,6alpha-epoxyergosta-8(14),22-dien-3beta-ol (1) and 22E-3beta-hydroxy-5alpha,6alpha,8alpha,14alpha-diepoxyergosta-22-en-7-one (2). The NMR data and complete assignments for both DMSO-d(6) and CDCl(3) were given. Their cytotoxic activity against A549 cell line was evaluated. Furthermore, the detailed conformation analysis for ring B (cyclohexene oxide system) of sterol 1 was given on the basis of NOEs. The endo-boat conformation was considered as the preferred conformation for ring B rather than half-chair conformation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C102504
Cyclohexene oxide, 98%
C6H10O