EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
The Henry reaction of (1R)-(1,4:3,6-dianhydro-D-mannitol-2-yl)-1,4:3,6-dianhydro-D-fructose 5,5'-dinitrate. Different reactive features of nitromethane to nitroethane.
PMID 19880098
Abstract
Henry reactions of a novel higher sugar derivative, (1R)-(1,4:3,6-dianhydro-D-mannitol-2-yl)-1,4:3,6-dianhydro-D-fructose 5,5'-dinitrate (Alternate nomenclature: (1R)-(isomannid-2-yl)-1,4:3,6-dianhydro-D-fructose 5,5'-dinitrate), with nitromethane and nitroethane were studied. The kinetic and thermodynamic reactions with nitromethane under different conditions were carried out to afford (2S)- and (2R)-beta-nitroalcohols, respectively. But when using nitroethane the reaction gave a (2S)-beta-nitroalcohol with an inverted configuration at vicinal carbon C-1. Two stereogenic centers were generated, and one was altered in the reaction.