EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
COX, LOX and platelet aggregation inhibitory properties of Lauraceae neolignans.
PMID 19880317
Abstract
The anti-inflammatory potential of 26 neolignans (14 of the bicyclooctane-type and 12 of the benzofuran-type), isolated from three Lauraceae species (Pleurothyrium cinereum, Ocotea macrophylla and Nectandra amazonum), was evaluated in vitro through inhibition of COX-1, COX-2, 5-LOX and agonist-induced aggregation of rabbit platelets. Benzofuran neolignans were found to be selective COX-2 inhibitors, whereas bicyclooctane neolignans inhibit selectively the PAF-action as well as COX-1 and 5-LOX. The neolignan 9-nor-7,8-dehydro-isolicarin B 15 and cinerin C 7 were found to be the most potent COX-2 inhibitor and PAF-antagonist, respectively. Nectamazin C 10 exhibited dual 5-LOX/COX-2 inhibition.