EMAIL THIS PAGE TO A FRIEND
Critical reviews in eukaryotic gene expression
A kinetic overview of the receptors involved in 1,25-dihydroxyvitamin D3 and 24,25-dihydroxyvitamin D3 signaling: a systems biology approach.
PMID 19883364
Abstract
The vitamin D endocrine system modulates an arsenal of important biological functions in more than 30 different tissues in short- and long-term perspectives. Two membrane receptors and one nuclear receptor are suggested to be involved in the vitamin D signaling system, but the function and physiological relevance of the receptors are debated. The complexity of the vitamin D endocrine system makes it necessary to combine experimental data with in silico simulations to get a holistic view of vitamin D-dependent regulation of tissue and cell physiology. This review focus on binding characteristics for the three putative vitamin D receptors and proposes a future systems biology approach including mathematical modeling that will be helpful together with experimental methods in depicting antitumoral and other biological effects promoted by the vitamin D endocrine system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17943
(24R)-24,25-Dihydroxyvitamin D3, ≥98% (vitamin + pre-vitamin, HPLC)
C27H44O3