EMAIL THIS PAGE TO A FRIEND
Cell Metabolism
Nesfatin-1-regulated oxytocinergic signaling in the paraventricular nucleus causes anorexia through a leptin-independent melanocortin pathway.
PMID 19883614