EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Molecular recognition of cationic phenothiazinium and phenoxazinium dyes with pi-extended 2'-deoxyadenosine nucleotides.
PMID 19885459
Abstract
N(6)-(N'-Arylcarbamoyl)-2'-deoxyadenosine-H-phosphonates displayed molecular recognition towards cationic phenothiazinium and phenoxazinium dyes in aqueous solutions; studies have shown that binding is driven mainly by aromatic interactions and that size and shape-complementarity of the aromatic rings in host and guest provides selectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
263893
Phenoxazine, ≥99%, purified by sublimation
C12H9NO
P14858
Phenoxazine, 97%
C12H9NO