EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Evidence for the formation of a mononuclear ferric-hydroperoxo complex via the reaction of dioxygen with an (N4S(thiolate))iron(II) complex.
PMID 19885493
Abstract
Addition of O(2)(g) at low-temperature to a mononuclear, nonheme iron(ii) complex comprising a tetraazamacrocyclic N(4) donor and an arylthiolate S donor leads to the generation of a deep red complex assigned as a low-spin Fe(III)?OOH complex, formed via metal- and ligand-assisted oxidation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W318000
2-Mercaptopropionic acid, ≥95%, FG
C3H6O2S
T31003
Thiolactic acid, 95%
C3H6O2S