EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Lewis base-catalyzed three-component Strecker reaction on water. An efficient manifold for the direct alpha-cyanoamination of ketones and aldehydes.
PMID 19885497
Abstract
The first three-component organocatalyzed Strecker reaction operating on water has been developed. The manifold utilizes ketones (aldehydes) as the starting carbonyl component, aniline as the primary amine, acetyl cyanide as the cyanide source and N,N-dimethylcyclohexylamine as the catalyst.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
290629
N,N-Dimethylcyclohexylamine, 99%
C8H17N