EMAIL THIS PAGE TO A FRIEND
Oncology reports
Triptolide induces apoptosis in human anaplastic thyroid carcinoma cells by a p53-independent but NF-kappaB-related mechanism.
PMID 19885592
Abstract
Triptolide, a traditional anti-inflammatory and anti-immunodepressive agent, has been reported to exert anti-neoplastic activity on several human tumor cell lines. This study investigates the pro-apoptotic function and the functional mechanism of triptolide on anaplastic thyroid carcinoma (ATC) cells. Experiments presented here demonstrated that triptolide had dose-dependent effects on cell viability of human ATC cell line TA-K cells through inducing cell apoptosis. In the molecular level, triptolide did not successfully initiate p53 signaling pathway, but downregulated the nuclear factor kappaB (NF-kappaB) pathway. Our studies suggest that triptolide functions as an effective apoptotic inducer in a p53-independent, but NF-kappaB-dependent mechanism, thus providing a promising agent for tumor types with p53 mutation/deletion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3652
Triptolide, from Tripterygium wilfordii, ≥98% (HPLC), solid
C20H24O6