EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Cell cycle deregulation by a poxvirus partial mimic of anaphase-promoting complex subunit 11.
PMID 19887645
Abstract
The anaphase-promoting complex (APC), or cyclosome, is a ubiquitin ligase with major roles in cell cycle regulation. It is required for mitotic exit, but must be deactivated for the G(1)/S phase transition to occur. APC consists of at least 12 subunits with the catalytic core formed by a scaffold protein, APC2, and a RING-H2 protein, APC11. APC11 facilitates ubiquitin chain formation by recruiting ubiquitin-charged conjugating enzymes through its RING-H2 domain. We report that a small number of poxviruses encode RING-H2 proteins with sequence similarities to APC11. We show that a representative of these viral proteins mimics APC11 in its interactions with APC, but unlike APC11, the viral protein fails to promote ubiquitin chain formation. This absence of ubiquitin ligase activity is linked to a distinctive sequence variation within its RING-H2 domain. Expression of the viral protein led to cell cycle deregulation and the accumulation of APC substrates in a manner consistent with impaired APC function. Our data characterize this protein as a regulator of APC activity, and consequently, we have called it PACR (poxvirus APC/cyclosome regulator). Deletion of the PACR gene substantially reduced viral replication. Here, we report a viral mimic of an APC component and reveal an intriguing mechanism by which viruses can manipulate cell cycle progression and, thereby, promote their own replication.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E3403 EZview Red Protein G Affinity Gel