EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
NMR studies of the conformation of the natural sweetener rebaudioside A.
PMID 19889398
Abstract
Rebaudioside A is a natural sweetener from Stevia rebaudiana in which four beta-D-glucopyranose units are attached to the aglycone steviol. Its (1)H and (13)C NMR spectra in pyridine-d(5) were assigned using 1D and 2D methods. Constrained molecular dynamics of solvated rebaudioside using NMR constraints derived from ROESY cross peaks yielded the orientation of the beta-D-glucopyranose units. Hydrogen bonding was examined using the temperature coefficients of the hydroxyl chemical shifts, ROESY and long-range COSY spectra, and proton-proton coupling constants.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01432
Rebaudioside A, ≥96% (HPLC)
C44H70O23