EMAIL THIS PAGE TO A FRIEND
Journal of virology
Adeno-associated virus type 5 utilizes alternative translation initiation to encode a small Rep40-like protein.
PMID 19889770
Abstract
Alternative splicing of adeno-associated virus type 2 (AAV2) P19-generated pre-mRNAs generates the small Rep proteins Rep52 and Rep40, which differ in their carboxyl termini. Both proteins are required for optimal packaging of AAV2 genomes. AAV5 Rep-encoding P19-generated transcripts are primarily polyadenylated within the central intron and not efficiently spliced; however, surprisingly, AAV5 was found to generate high levels of a Rep40-like protein. The AAV5 Rep40-like protein was generated by internal initiation and has the same C terminus as Rep52. Although precluded from using alternative splicing to generate multiple Rep isoforms, AAV5 ensures the production of a Rep40-like protein by utilizing a novel internal translation initiation event.