EMAIL THIS PAGE TO A FRIEND
Hypertension in pregnancy
Digitalis-like induced arrhythmia in a patient with rheumatic mitral regurgitation complicated by preeclampsia.
PMID 19891530
Abstract
To report a case of cardiac arrhythmia related to a low dose of endovenous lanatoside C. A 23-year-old pregnant woman with mitral regurgitation complicated with preeclampsia and pulmonary edema presented 2 episodes of atrial tachycardia induced by a intravenous digitalis (2 mg, IV and 1 mg, IV, respectively). This case calls attention to the need for further studies analysing the security of digoxin use in preeclampsia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2261
Lanatoside C, ≥95%
C49H76O20