EMAIL THIS PAGE TO A FRIEND
Circulation Research
MicroRNA-133a protects against myocardial fibrosis and modulates electrical repolarization without affecting hypertrophy in pressure-overloaded adult hearts.
PMID 19893015
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-491 Anti-Kv4.2 Antibody