EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Short-step synthesis of a resveratrol derivative from commercially available 1,3-dimethoxybenzene and 4-vinylanisole.
PMID 19897904
Abstract
An efficient synthesis of tri-O-methylated resveratrol is presented using an advanced Heck reaction promoted by Pd(dba)(2) in the presence of P(t-Bu)(3).
Related Materials