EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Recovery of succinic acid from fermentation broth.
PMID 19898782
Abstract
Succinic acid is of high interest as bio-feedstock for the chemical industry. It is a precursor for a variety of many other chemicals, e.g. 1,4-butandiol, tetrahydrofuran, biodegradable polymers and fumaric acid. Besides optimized production strains and fermentation processes it is indispensable to develop cost-saving and energy-effective downstream processes to compete with the current petrochemical production process. Various methods such as precipitation, sorption and ion exchange, electrodialysis, and liquid-liquid extraction have been investigated for the recovery of succinic acid from fermentation broth and are reviewed critically here.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1418
Succinic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
C4H6O4