EMAIL THIS PAGE TO A FRIEND
Molecular diversity
Chloramine-T and N-chlorosuccinimide: novel reagents for diastereoselective oxidation of bisnaphthols.
PMID 19898947
Abstract
Two N-chloro reagents (chloramine-T and N-chlorosuccinimide) were used as effective oxidizing agents for the oxidation of bisnaphthols to the corresponding Abel's ketones under mild and convenient conditions at room temperature with good to excellent yields.