EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
An in vivo fluorescent sensor reveals intracellular ins(1,3,4,5)P4 dynamics in single cells.
PMID 19899175
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I8636
D-myo-Inositol 1,3,4,5-tetrakis(phosphate) ammonium salt, ≥95%
C6H16O18P4·8H3N