EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Lamiridosins, hepatitis C virus entry inhibitors from Lamium album.
PMID 19904996
Abstract
Phytochemical study of the aqueous extract of the flowering tops of Lamium album led to identification of the antiviral iridoid isomers lamiridosins A and B (1, 2). These compounds were found to significantly inhibit hepatitis C virus entry (IC(50) 2.31 muM) in vitro. Studies of 14 iridoid analogues showed that, while the parent iridoid glucosides demonstrated no anti-HCV entry activity, the aglycones of shanzhiside methyl ester (4), loganin (5), loganic acid (6), geniposide (10), verbenalin (12), eurostoside (15), and picroside II (17) exhibited significant anti-HCV entry and anti-infectivity activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00231
Loganic acid, ≥95% (LC/MS-ELSD)
C16H24O10
36483
Loganin, analytical standard
C17H26O10