EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Role of the serotoninergic system in the development of diseases of the intestinal and biliferous tracts.
PMID 19907762
Abstract
Primary sclerosing cholangitis and nonspecific ulcerative colitis were modeled before and after serotonin and spiperone injections. Activation of the serotoninergic system prevented the development of primary sclerosing cholangitis, but promoted more severe course of nonspecific ulcerative colitis.