EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthetic routes to pyrrolizine-1,5-dione derivatives by flash vacuum pyrolysis of amidomethylene derivatives of Meldrum's acid.
PMID 19907784
Abstract
Methoxymethylene Meldrum's acid 1 reacts with 5- and 6-membered lactams in refluxing acetonitrile to give the N-substituted products 9-15. If the reactions are continued for extended times, the Meldrum's acid derivatives decompose to provide enamidoesters e.g. 22-24. Flash vacuum pyrolysis of the 5-membered ring products 9-13 provides reasonable yields of the fused pyrrolones 31-35. The constitution of the products is supported by X-ray crystal structures of 10, 12, 19, 32 and 34.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
210145
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
C6H8O4