EMAIL THIS PAGE TO A FRIEND
American journal of medical genetics. Part A
Autosomal dominant syndrome of mental retardation, hypotelorism, and cleft palate resembling Schilbach-Rott syndrome.
PMID 19921646
Abstract
We present a family segregating for an autosomal dominant syndrome of hypotelorism, cleft palate/uvula, high-arched palate and mild mental retardation. Although these findings may suggest a form of holoprosencephaly, no holoprosencephaly was found on MRI of the proposita. Results of genetic studies were normal including FISH for deletion of 22q11, karyotype analysis, fragile X testing, high-resolution comparative genomic hybridization and SEPT9, SHH mutation analysis. The syndrome is reminiscent of the infrequently recognized autosomal dominant Schilbach-Rott syndrome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5050 Sephadex® G-50, Superfine