EMAIL THIS PAGE TO A FRIEND
Clinical and experimental dermatology
Etretinate combined with low-dose prednisolone for an aged patient with adult T-cell leukaemia/lymphoma.
PMID 19925487
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1011029 Acitretin Related Compound B, United States Pharmacopeia (USP) Reference Standard
C23H30O3