EMAIL THIS PAGE TO A FRIEND
British journal of haematology
A unified definition of clinical resistance and intolerance to hydroxycarbamide in polycythaemia vera and primary myelofibrosis: results of a European LeukemiaNet (ELN) consensus process.
PMID 19930182