EMAIL THIS PAGE TO A FRIEND
American journal of therapeutics
Effect of laropiprant, a PGD2 receptor 1 antagonist, on estradiol and norgestimate pharmacokinetics after oral contraceptive administration in women.
PMID 19940609
Abstract
Laropiprant is a prostaglandin D2 receptor 1 antagonist that is being developed in combination with niacin for the treatment of dyslipidemia. This randomized clinical study evaluated the effect of laropiprant on the pharmacokinetics of ethinyl estradiol (EE) and norelgestromin (NGMN), the principal circulating metabolite of norgestimate, in healthy women receiving 3 or more months of an oral contraceptive (Ortho Tri-Cyclen; Ortho-McNeil Pharmaceutical, Raritan, NJ), which contains EE and norgestimate. Twenty-one female subjects with normal menstrual cycles received the oral contraceptive on Days 1 to 21 during two consecutive contraceptive cycles. Subjects received double-blind 40 mg/day laropiprant or placebo on Days 1 to 21 of each contraceptive cycle. Plasma samples were collected predose and 0.5, 1, 1.5, 2, 3, 4, 6, 8, 12, and 24 hours postdose on Day 21 to measure area under the plasma concentration-time curve from 0 to 24 hours (AUC0-24hr) and maximum concentration observed in plasma (Cmax) of EE and NGMN. Comparability would be declared if the 90% confidence intervals for the geometric mean ratio of AUC0-24hr and Cmax in the absence and presence of laropiprant were within predefined bounds (0.80-1.25). The estimated geometric mean ratios (90% confidence intervals) of EE and NGMN, respectively, were 1.08 (1.04-1.13) and 0.97 (0.94-0.99) for AUC0-24hr and 1.16 (1.06-1.27) and 1.00 (0.94-1.06) for Cmax. The 90% confidence intervals for the geometric mean ratio of EE Cmax minimally exceeded the prespecified bounds; the other relevant pharmacokinetic parameters fell within the predefined bounds. Coadministration of 40 mg laropiprant with the oral contraceptive did not lead to clinically meaningful alterations in the pharmacokinetics of EE or NGMN.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94497
Norgestimate, analytical standard
C23H31NO3
SML0359
Norgestimate, ≥97% (HPLC)
C23H31NO3